accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 1074-82-4

IUPAC Name: potassium 1,3-dioxo-2,3-dihydro-1H-isoindol-2-ide
Synonyms: potassium phthalimide
Molecular Weight (g/mol): 185.22
Molecular Formula: C8H4KNO2
InChi Key: FYRHIOVKTDQVFC-UHFFFAOYSA-M
SMILES: [K+].O=C1[N-]C(=O)C2=CC=CC=C12
Quantity 
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
Percent Purity 
  • (3)
  • (3)
Physical Form 
  • (3)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (3)
  • (3)
  • (1)