accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

FFOHO

CAS RN 107496-54-8

IUPAC Name: 3,3-difluorocyclobutane-1-carboxylic acid
Synonyms: 3,3-difluorocyclobutane-1-carboxylic acid
Molecular Weight (g/mol): 136.1
Molecular Formula: C5H6F2O2
InChi Key: PLRCVBKYFLWAAT-UHFFFAOYSA-N
SMILES: OC(=O)C1CC(F)(F)C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results