accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 1075-35-0

IUPAC Name: 5-chloro-2-methyl-1H-indole
Synonyms: 5-chloro-2-methyl-1H-indole
Molecular Weight (g/mol): 165.62
Molecular Formula: C9H8ClN
InChi Key: WUVWAXJXPRYUME-UHFFFAOYSA-N
SMILES: CC1=CC2=CC(Cl)=CC=C2N1
Percent Purity 
  • (1)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)