accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 1075-82-7

CH3OOOH

CAS RN 1075-82-7

IUPAC Name: ethyl 1-(hydroxymethyl)cyclopentane-1-carboxylate
Synonyms: ethyl 1-(hydroxymethyl)cyclopentane-1-carboxylate
Molecular Weight (g/mol): 172.22
Molecular Formula: C9H16O3
InChi Key: AUUICUZWYQYTNO-UHFFFAOYSA-N
SMILES: CCOC(=O)C1(CO)CCCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results