accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

HOOO

CAS RN 1076-38-6

IUPAC Name: 2-hydroxy-4H-chromen-4-one
Synonyms: 4-hydroxycoumarin 3-(4-hydroxyphenyl)-1-propane benzotetronic acid 3-ohpp 4-hydroxy-1-benzopyran-2-one 4-coumaryl alcohol hydroxy coumarin p-coumaryl alcohol benzotetronate
Molecular Weight (g/mol): 162.14
Molecular Formula: C9H6O3
InChi Key: OWBBAPRUYLEWRR-UHFFFAOYSA-N
SMILES: OC1=CC(=O)C2=CC=CC=C2O1
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
Percent Purity 
  • (3)
  • (3)
Certifications/Compliance 
  • (3)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results