accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 1077-16-3

H3C

CAS RN 1077-16-3

IUPAC Name: hexylbenzene
Synonyms: N-hexylbenzene
Molecular Weight (g/mol): 162.28
Molecular Formula: C12H18
InChi Key: LTEQMZWBSYACLV-UHFFFAOYSA-N
SMILES: CCCCCCC1=CC=CC=C1
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)