accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

BrNHNOOH

CAS RN 1077-94-7

IUPAC Name: 5-bromo-1H-indazole-3-carboxylic acid
Synonyms: 5-bromo-1H-indazole-3-carboxylic acid
Molecular Weight (g/mol): 241.04
Molecular Formula: C8H5BrN2O2
InChi Key: AMJVXOOGGBPVCZ-UHFFFAOYSA-N
SMILES: OC(=O)C1=NNC2=CC=C(Br)C=C12
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results