accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NH2SNHClCl

CAS RN 107707-33-5

IUPAC Name: (3,5-dichlorophenyl)thiourea
Synonyms: 3,5-dichlorophenylthiourea
Molecular Weight (g/mol): 221.1
Molecular Formula: C7H6Cl2N2S
InChi Key: BNJARXOEVLYKDN-UHFFFAOYSA-N
SMILES: NC(=S)NC1=CC(Cl)=CC(Cl)=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results