CAS RN 107754-20-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
4-[[5-[[(Cyclopentyloxy)carbonyl]amino]-1-methyl-1H-indol-3-yl]methyl]-3-methoxybenzoic Acid, TRC
CAS: 107754-20-1 Molecular Formula: C24 H26 N2 O5 Molecular Weight (g/mol): 422.47 Synonym: Benzoic acid, 4-[[5-[[(cyclopentyloxy)carbonyl]amino]-1-methyl-1H-indol-3-yl]methyl]-3-methoxy-,4-[[5-[[(Cyclopentyloxy)carbonyl]amino]-1-methyl-1H-indol-3-yl]methyl]-3-methoxybenzoic acid IUPAC Name: 4-[[5-(cyclopentyloxycarbonylamino)-1-methylindol-3-yl]methyl]-3-methoxybenzoic acid SMILES: COc1cc(ccc1Cc2cn(C)c3ccc(NC(=O)OC4CCCC4)cc23)C(=O)O