accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

ClP

CAS RN 1079-66-9

IUPAC Name: chlorodiphenylphosphane
Synonyms: chlorodiphenylphosphine
Molecular Weight (g/mol): 220.64
Molecular Formula: C12H10ClP
InChi Key: XGRJZXREYAXTGV-UHFFFAOYSA-N
SMILES: ClP(C1=CC=CC=C1)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (3)
  • (13)
Quantity 
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (3)
  • (1)
  • (1)
Boiling Point 
  • (6)
  • (7)
Percent Purity 
  • (3)
  • (3)
  • (3)
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (6)
  • (6)
  • (3)
  • (13)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (3)
  • (1)
  • (1)
  • (6)
  • (7)
  • (3)
  • (3)
  • (3)
  • (4)
  • (7)
  • (3)