CAS RN 108-18-9
Diisopropylamine, 99+%, Thermo Scientific Chemicals
CAS: 108-18-9 Molecular Weight (g/mol): 101.19 MDL Number: MFCD00008862 InChI Key: UAOMVDZJSHZZME-UHFFFAOYSA-N Synonym: diisopropylamine,2-propanamine, n-1-methylethyl,n-1-methylethyl-2-propanamine,dipa,n,n-diisopropylamine,bis propan-2-yl amine,n-isopropylpropan-2-amine,di-isopropylamine,n-isopropyl-1-amino-2-methylethane,diisopropyl amine PubChem CID: 7912 SMILES: CC(C)NC(C)C
Diisopropylamine, 99.5%, redistilled, AcroSeal™, Thermo Scientific Chemicals
CAS: 108-18-9 Molecular Formula: C6H15N Molecular Weight (g/mol): 101.19 MDL Number: MFCD00008862 InChI Key: UAOMVDZJSHZZME-UHFFFAOYSA-N Synonym: diisopropylamine,2-propanamine, n-1-methylethyl,n-1-methylethyl-2-propanamine,dipa,n,n-diisopropylamine,bis propan-2-yl amine,n-isopropylpropan-2-amine,di-isopropylamine,n-isopropyl-1-amino-2-methylethane,diisopropyl amine PubChem CID: 7912 IUPAC Name: N-propan-2-ylpropan-2-amine SMILES: CC(C)NC(C)C
Diisopropylamine, 99%, Thermo Scientific Chemicals
CAS: 108-18-9 InChI Key: UAOMVDZJSHZZME-UHFFFAOYSA-N Synonym: diisopropylamine,2-propanamine, n-1-methylethyl,n-1-methylethyl-2-propanamine,dipa,n,n-diisopropylamine,bis propan-2-yl amine,n-isopropylpropan-2-amine,di-isopropylamine,n-isopropyl-1-amino-2-methylethane,diisopropyl amine PubChem CID: 7912 IUPAC Name: N-propan-2-ylpropan-2-amine SMILES: CC(C)NC(C)C
Diisopropylamine, 99+%, Thermo Scientific Chemicals
CAS: 108-18-9 Molecular Formula: C6H15N Molecular Weight (g/mol): 101.193 MDL Number: MFCD00008862 InChI Key: UAOMVDZJSHZZME-UHFFFAOYSA-N Synonym: diisopropylamine,2-propanamine, n-1-methylethyl,n-1-methylethyl-2-propanamine,dipa,n,n-diisopropylamine,bis propan-2-yl amine,n-isopropylpropan-2-amine,di-isopropylamine,n-isopropyl-1-amino-2-methylethane,diisopropyl amine PubChem CID: 7912 IUPAC Name: N-propan-2-ylpropan-2-amine SMILES: CC(C)NC(C)C