accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NH2ONHNH2O

CAS RN 108-19-0

IUPAC Name: 1-(carbamoylamino)formamide
Synonyms: biuret epa pesticide chemical code 206200 2-imidodicarbonic diamide allophanamide isobiuret allophanimidic acid caswell no. 159A caswell no. 106A ureidoformamide imidodicarbonic diamide
Molecular Weight (g/mol): 103.08
Molecular Formula: C2H5N3O2
InChi Key: OHJMTUPIZMNBFR-UHFFFAOYSA-N
SMILES: NC(=O)NC(N)=O
Molecular Weight (g/mol) 
  • (6)
  • (2)
Quantity 
  • (2)
  • (2)
  • (2)
  • (1)
Grade 
  • (3)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results