CAS RN 108-32-7
Propylene carbonate, 99%
CAS: 108-32-7 Molecular Formula: C4H6O3 Molecular Weight (g/mol): 102.09 MDL Number: MFCD00005385,MFCD00798264,MFCD00798265 InChI Key: RUOJZAUFBMNUDX-UHFFFAOYNA-N Synonym: propylene carbonate,1,2-propylene carbonate,1,2-propanediol cyclic carbonate,texacar pc,1,3-dioxolan-2-one, 4-methyl,cyclic propylene carbonate,arconate 5000,1,2-propanediol carbonate,1-methylethylene carbonate,cyclic 1,2-propylene carbonate PubChem CID: 7924 IUPAC Name: 4-methyl-1,3-dioxolan-2-one SMILES: CC1COC(=O)O1
Propylene carbonate, 99.5%
CAS: 108-32-7 Molecular Formula: C4H6O3 Molecular Weight (g/mol): 102.09 MDL Number: MFCD00005385,MFCD00798264,MFCD00798265 InChI Key: RUOJZAUFBMNUDX-UHFFFAOYNA-N Synonym: propylene carbonate,1,2-propylene carbonate,1,2-propanediol cyclic carbonate,texacar pc,1,3-dioxolan-2-one, 4-methyl,cyclic propylene carbonate,arconate 5000,1,2-propanediol carbonate,1-methylethylene carbonate,cyclic 1,2-propylene carbonate PubChem CID: 7924 IUPAC Name: 4-methyl-1,3-dioxolan-2-one SMILES: CC1COC(=O)O1
Propylene carbonate, 99.5%, anhydrous, AcroSeal™
CAS: 108-32-7 Molecular Formula: C4H6O3 Molecular Weight (g/mol): 102.09 MDL Number: MFCD00005385,MFCD00798264,MFCD00798265 InChI Key: RUOJZAUFBMNUDX-UHFFFAOYNA-N Synonym: propylene carbonate,1,2-propylene carbonate,1,2-propanediol cyclic carbonate,texacar pc,1,3-dioxolan-2-one, 4-methyl,cyclic propylene carbonate,arconate 5000,1,2-propanediol carbonate,1-methylethylene carbonate,cyclic 1,2-propylene carbonate PubChem CID: 7924 IUPAC Name: 4-methyl-1,3-dioxolan-2-one SMILES: CC1COC(=O)O1
Propylene carbonate, MP Biomedicals™
CAS: 108-32-7 Molecular Formula: C4H6O3 Molecular Weight (g/mol): 102.09 MDL Number: MFCD00005385,MFCD00798264,MFCD00798265 InChI Key: RUOJZAUFBMNUDX-UHFFFAOYNA-N Synonym: propylene carbonate,1,2-propylene carbonate,1,2-propanediol cyclic carbonate,texacar pc,1,3-dioxolan-2-one, 4-methyl,cyclic propylene carbonate,arconate 5000,1,2-propanediol carbonate,1-methylethylene carbonate,cyclic 1,2-propylene carbonate PubChem CID: 7924 IUPAC Name: 4-methyl-1,3-dioxolan-2-one SMILES: CC1COC(=O)O1
Propylene Carbonate, TRC
CAS: 108-32-7 Molecular Formula: C4 H6 O3 Molecular Weight (g/mol): 102.09 Synonym: 1,3-Dioxolan-2-one, 4-methyl-,Carbonic acid, cyclic propylene ester (8CI),Carbonic acid, propylene ester (6CI),4-Methyl-1,3-dioxolan-2-one,1,2-Propanediol carbonate,1,2-Propanediol cyclic carbonate,1,2-Propanediyl carbonate,1,2-Propylene carbonate,1-Methylethylene carbonate,2-Methyl-1,2-ethylene carbonate,2-Oxo-4-methyl-1,3-dioxolane,4-Methyl-1,3-dioxolane-2-one,4-Methyl-2-dioxolanone,4-Methyl-2-oxo-1,3-dioxolane,Arconate 1000,Arconate 5000,Arconate HP,Arconate propylenecarbonate,Carbonic acid cyclic 1,2-propylene ester,Carbonic acid cyclic methylethylene ester,Cyclic 1,2-propylene carbonate,Cyclic methylethylene carbonate,Cyclic propylene carbonate,Fluor Solvent,JN 02,Jeffsol 1555,Jeffsol AG 1555,Jeffsol PC,Methyl-1,3-dioxolan-2-one,NSC 11784,NSC 1913,PC Medion,PC-HP,Propylene carbonate,Propylene glycol cyclic carbonate,Texacar PC IUPAC Name: 4-methyl-1,3-dioxolan-2-one SMILES: CC1COC(=O)O1
Propylene Carbonate, 99%, Reagent Grade, Honeywell™
CAS: 108-32-7 Molecular Formula: C4H6O3 Molecular Weight (g/mol): 102.09 MDL Number: MFCD00005385,MFCD00798264,MFCD00798265 InChI Key: RUOJZAUFBMNUDX-UHFFFAOYNA-N Synonym: propylene carbonate,1,2-propylene carbonate,1,2-propanediol cyclic carbonate,texacar pc,1,3-dioxolan-2-one, 4-methyl,cyclic propylene carbonate,arconate 5000,1,2-propanediol carbonate,1-methylethylene carbonate,cyclic 1,2-propylene carbonate PubChem CID: 7924 IUPAC Name: 4-methyl-1,3-dioxolan-2-one SMILES: CC1COC(=O)O1
HYDRANAL™ - Water Standard 10.0, Standard for Karl Fischer titration (water content 10 mg/g = 1.0%), Solstice
Molecular Formula: C8H10 Molecular Weight (g/mol): 106.17 MDL Number: MFCD00008519 InChI Key: CTQNGGLPUBDAKN-UHFFFAOYSA-N Synonym: 1,2-Propanediol cyclic carbonate,4-Methyl-1,3-dioxolan-2-one PubChem CID: 962 ChEBI: CHEBI:15377 IUPAC Name: 1,2-xylene SMILES: CC1=CC=CC=C1C