accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3H3C

CAS RN 108-38-3

IUPAC Name: 1,3-xylene
Synonyms: M-xylene m-xylol ccris 907
Molecular Weight (g/mol): 106.17
Molecular Formula: C8H10
InChi Key: IVSZLXZYQVIEFR-UHFFFAOYSA-N
SMILES: CC1=CC(C)=CC=C1
Grade 
  • (2)
  • (4)
Percent Purity 
  • (4)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
Solvent or Matrix 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (1)
  • (6)
  • (1)
  • (2)
  • (4)
  • (4)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)