accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 108-39-4

IUPAC Name: 3-methylphenol
Synonyms: M-cresol fema number 2337 rover dog shampoo cresol m-kresol celcure dry mix (chemicals for wood preserving) m-toluol epa pesticide chemical code 022102 ccris 645 rcra waste number U052
Molecular Weight (g/mol): 108.14
Molecular Formula: C7H8O
InChi Key: RLSSMJSEOOYNOY-UHFFFAOYSA-N
SMILES: CC1=CC=CC(O)=C1
Percent Purity 
  • (2)
  • (4)
  • (7)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (3)
  • (2)
  • (1)
  • (1)
Boiling Point 
  • (4)
  • (5)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (4)
  • (1)
  • (1)
  • (1)
  • (5)
  • (2)
  • (2)
  • (2)
  • (4)
  • (7)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (3)
  • (2)
  • (1)
  • (1)
  • (4)
  • (5)
  • (9)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)