accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 108-53-2

IUPAC Name: 2-amino-3,4-dihydropyrimidin-4-one
Synonyms: isocytosine
Molecular Weight (g/mol): 111.1
Molecular Formula: C4H5N3O
InChi Key: XQCZBXHVTFVIFE-UHFFFAOYSA-N
SMILES: NC1=NC=CC(=O)N1
Molecular Weight (g/mol) 
  • (1)
  • (5)
Quantity 
  • (1)
  • (2)
  • (3)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (5)
  • (1)
  • (2)
  • (3)
  • (2)