accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3OOOH3CO

CAS RN 108-58-7

IUPAC Name: 3-(acetyloxy)phenyl acetate
Synonyms: 1,3-benzenediol, diacetate
Molecular Weight (g/mol): 194.19
Molecular Formula: C10H10O4
InChi Key: STOUHHBZBQBYHH-UHFFFAOYSA-N
SMILES: CC(=O)OC1=CC(OC(C)=O)=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results