accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NH2NNH2NH2NN

CAS RN 108-78-1

IUPAC Name: 1,3,5-triazine-2,4,6-triamine
Synonyms: melamine pluragard adk stab ZS 27 yukamelamine melaminium citrate mark ZS 27 s-triazinetriamine teoharn cyanuramide theoharn
Molecular Weight (g/mol): 126.12
Molecular Formula: C3H6N6
InChi Key: JDSHMPZPIAZGSV-UHFFFAOYSA-N
SMILES: NC1=NC(N)=NC(N)=N1
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
Percent Purity 
  • (3)
  • (7)
Certifications/Compliance 
  • (2)
  • (4)
  • (1)
Physical Form 
  • (4)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (2)
  • (4)
  • (3)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (3)
  • (7)
  • (2)
  • (4)
  • (1)
  • (4)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)