accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CCH3OCH3CH3

CAS RN 108-83-8

IUPAC Name: 2,6-dimethylheptan-4-one
Synonyms: 2,6-dimethyl-4-heptanone
Molecular Weight (g/mol): 142.24
Molecular Formula: C9H18O
InChi Key: PTTPXKJBFFKCEK-UHFFFAOYSA-N
SMILES: CC(C)CC(=O)CC(C)C
Boiling Point 
  • (3)
  • (3)
Molecular Weight (g/mol) 
  • (3)
  • (3)
Percent Purity 
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Grade 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (3)