accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

Br

CAS RN 108-86-1

IUPAC Name: bromobenzene
Synonyms: bromobenzene bromobenzene (substance) bromobenzene, 14C-labeled 14c-labeled bromobenzene 14C-labeled bromobenzene PhBr bromobenzene, 14c-labeled C6H5br hsdb 47
Molecular Weight (g/mol): 157.01
Molecular Formula: C6H5Br
InChi Key: QARVLSVVCXYDNA-UHFFFAOYSA-N
SMILES: BrC1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (3)
  • (7)
Boiling Point 
  • (3)
  • (1)
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (4)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)
  • (7)
  • (3)
  • (1)
  • (4)
  • (5)
  • (4)
  • (1)