CAS RN 108-90-7
Chlorobenzene, 99+%, pure
CAS: 108-90-7 Molecular Formula: C6H5Cl Molecular Weight (g/mol): 112.556 MDL Number: MFCD00000530 InChI Key: MVPPADPHJFYWMZ-UHFFFAOYSA-N Synonym: monochlorobenzene,benzene chloride,phenyl chloride,benzene, chloro,chlorbenzene,chlorobenzol,monochlorbenzol,chlorbenzol,chlorobenzen,clorobenzene PubChem CID: 7964 ChEBI: CHEBI:28097 IUPAC Name: chlorobenzene SMILES: C1=CC=C(C=C1)Cl
Chlorobenzene, Certified AR for Analysis, Fisher Chemical™
CAS: 108-90-7 Molecular Formula: C6H5Cl Molecular Weight (g/mol): 112.56 MDL Number: 530 InChI Key: MVPPADPHJFYWMZ-UHFFFAOYSA-N IUPAC Name: chlorobenzene SMILES: ClC1=CC=CC=C1
Chlorobenzene, 99.8%, Extra Dry, AcroSeal™
CAS: 108-90-7 Molecular Formula: C6H5Cl Molecular Weight (g/mol): 112.556 MDL Number: MFCD00000530 InChI Key: MVPPADPHJFYWMZ-UHFFFAOYSA-N Synonym: monochlorobenzene,benzene chloride,phenyl chloride,benzene, chloro,chlorbenzene,chlorobenzol,monochlorbenzol,chlorbenzol,chlorobenzen,clorobenzene PubChem CID: 7964 ChEBI: CHEBI:28097 IUPAC Name: chlorobenzene SMILES: C1=CC=C(C=C1)Cl
Chlorobenzene, Extra Pure, SLR, Fisher Chemical™
CAS: 108-90-7 Molecular Formula: C6H5Cl Molecular Weight (g/mol): 112.56 MDL Number: 530 InChI Key: MVPPADPHJFYWMZ-UHFFFAOYSA-N IUPAC Name: chlorobenzene SMILES: ClC1=CC=CC=C1
Chlorobenzene, 99%
CAS: 108-90-7 Molecular Formula: C6H5Cl Molecular Weight (g/mol): 112.556 MDL Number: MFCD00000530 InChI Key: MVPPADPHJFYWMZ-UHFFFAOYSA-N Synonym: monochlorobenzene,benzene chloride,phenyl chloride,benzene, chloro,chlorbenzene,chlorobenzol,monochlorbenzol,chlorbenzol,chlorobenzen,clorobenzene PubChem CID: 7964 ChEBI: CHEBI:28097 IUPAC Name: chlorobenzene SMILES: C1=CC=C(C=C1)Cl
Chlorobenzene, 99.5%, Extra Dry over Molecular Sieve, AcroSeal™
CAS: 108-90-7 Molecular Formula: C6H5Cl Molecular Weight (g/mol): 112.556 InChI Key: MVPPADPHJFYWMZ-UHFFFAOYSA-N Synonym: monochlorobenzene,benzene chloride,phenyl chloride,benzene, chloro,chlorbenzene,chlorobenzol,monochlorbenzol,chlorbenzol,chlorobenzen,clorobenzene PubChem CID: 7964 ChEBI: CHEBI:28097 IUPAC Name: chlorobenzene SMILES: C1=CC=C(C=C1)Cl
Chlorobenzene, 99.9%, for HPLC
CAS: 108-90-7 Molecular Formula: C6H5Cl Molecular Weight (g/mol): 112.56 InChI Key: MVPPADPHJFYWMZ-UHFFFAOYSA-N Synonym: monochlorobenzene,benzene chloride,phenyl chloride,benzene, chloro,chlorbenzene,chlorobenzol,monochlorbenzol,chlorbenzol,chlorobenzen,clorobenzene PubChem CID: 7964 ChEBI: CHEBI:28097 IUPAC Name: chlorobenzene SMILES: C1=CC=C(C=C1)Cl
Chlorobenzene, ≥99.5%, ACS reagent
CAS: 108-90-7 Molecular Formula: C6H5Cl Molecular Weight (g/mol): 112.556 MDL Number: MFCD00000530 InChI Key: MVPPADPHJFYWMZ-UHFFFAOYSA-N Synonym: monochlorobenzene,benzene chloride,phenyl chloride,benzene, chloro,chlorbenzene,chlorobenzol,monochlorbenzol,chlorbenzol,chlorobenzen,clorobenzene PubChem CID: 7964 ChEBI: CHEBI:28097 IUPAC Name: chlorobenzene SMILES: C1=CC=C(C=C1)Cl
Chlorobenzene, Spectrophotometric Grade, 99.9%
CAS: 108-90-7 Molecular Formula: C6H5Cl Molecular Weight (g/mol): 112.556 MDL Number: MFCD00000530 InChI Key: MVPPADPHJFYWMZ-UHFFFAOYSA-N Synonym: monochlorobenzene,benzene chloride,phenyl chloride,benzene, chloro,chlorbenzene,chlorobenzol,monochlorbenzol,chlorbenzol,chlorobenzen,clorobenzene PubChem CID: 7964 ChEBI: CHEBI:28097 IUPAC Name: chlorobenzene SMILES: C1=CC=C(C=C1)Cl
Chlorobenzene, 99+%, for spectroscopy
CAS: 108-90-7 Molecular Formula: C6H5Cl Molecular Weight (g/mol): 112.556 MDL Number: MFCD00000530 InChI Key: MVPPADPHJFYWMZ-UHFFFAOYSA-N Synonym: monochlorobenzene,benzene chloride,phenyl chloride,benzene, chloro,chlorbenzene,chlorobenzol,monochlorbenzol,chlorbenzol,chlorobenzen,clorobenzene PubChem CID: 7964 ChEBI: CHEBI:28097 IUPAC Name: chlorobenzene SMILES: C1=CC=C(C=C1)Cl
Chlorobenzene, HPLC Grade, 99.5%
CAS: 108-90-7 Molecular Formula: C6H5Cl Molecular Weight (g/mol): 112.556 MDL Number: MFCD00000530 InChI Key: MVPPADPHJFYWMZ-UHFFFAOYSA-N Synonym: monochlorobenzene,benzene chloride,phenyl chloride,benzene, chloro,chlorbenzene,chlorobenzol,monochlorbenzol,chlorbenzol,chlorobenzen,clorobenzene PubChem CID: 7964 ChEBI: CHEBI:28097 IUPAC Name: chlorobenzene SMILES: C1=CC=C(C=C1)Cl
Chlorobenzene, HPLC Grade, 99+%
CAS: 108-90-7 | C{6}H{5}Cl
Chlorobenzene, ACS, 99.5%
CAS: 108-90-7 Molecular Formula: C6H5Cl Molecular Weight (g/mol): 112.556 MDL Number: MFCD00000530 InChI Key: MVPPADPHJFYWMZ-UHFFFAOYSA-N Synonym: monochlorobenzene,benzene chloride,phenyl chloride,benzene, chloro,chlorbenzene,chlorobenzol,monochlorbenzol,chlorbenzol,chlorobenzen,clorobenzene PubChem CID: 7964 ChEBI: CHEBI:28097 IUPAC Name: chlorobenzene SMILES: C1=CC=C(C=C1)Cl
Chlorobenzene, ReagentPlus™, 99%, Solstice
CAS: 108-90-7 Molecular Formula: C6H5Cl Molecular Weight (g/mol): 112.556 MDL Number: MFCD00000530 InChI Key: MVPPADPHJFYWMZ-UHFFFAOYSA-N Synonym: monochlorobenzene,benzene chloride,phenyl chloride,benzene, chloro,chlorbenzene,chlorobenzol,monochlorbenzol,chlorbenzol,chlorobenzen,clorobenzene PubChem CID: 7964 ChEBI: CHEBI:28097 IUPAC Name: chlorobenzene SMILES: C1=CC=C(C=C1)Cl
- 1
- 2