accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 108-98-5

IUPAC Name: benzenethiol
Synonyms: thiophenol rcra waste number P014 thiofenol usaf XR-19
Molecular Weight (g/mol): 110.17
Molecular Formula: C6H6S
InChi Key: RMVRSNDYEFQCLF-UHFFFAOYSA-N
SMILES: SC1=CC=CC=C1
Molecular Weight (g/mol) 
  • (4)
  • (6)
Percent Purity 
  • (2)
  • (3)
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (4)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (4)
  • (6)
  • (2)
  • (3)
  • (4)
  • (9)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (4)
  • (3)
  • (1)