CAS RN 108050-82-4
IUPAC Name:
3-[4-(4-phenyl-1,2,3,6-tetrahydropyridin-1-yl)butyl]-1H-indol-5-ol hydrochloride
Synonyms:
roxindole hydrochloride
Molecular Weight (g/mol):
382.93
Molecular Formula:
C23H27ClN2O
InChi Key:
ZCEPVNSWLLJECX-UHFFFAOYSA-N
SMILES:
Cl.OC1=CC=C2NC=C(CCCCN3CCC(=CC3)C3=CC=CC=C3)C2=C1
1
–
1
of
1
results
1
Roxindole Hydrochloride, TRC
CAS: 108050-82-4 Molecular Formula: C23H26N2O . (HCl) Molecular Weight (g/mol): 382.9 Synonym: 3-[4-(3,6-Dihydro-4-phenyl-1(2H)-pyridinyl)butyl]-1H-indol-5-ol Hydrochloride,3-[4-(3,6-dihydro-4-phenyl-1(2H)-pyridinyl)butyl]-1H-Indol-5-ol Monohydrochloride,Roxindole Hydrochloride IUPAC Name: 3-[4-(4-phenyl-3,6-dihydro-2H-pyridin-1-yl)butyl]-1H-indol-5-ol;hydrochloride SMILES: Cl.Oc1ccc2[nH]cc(CCCCN3CCC(=CC3)c4ccccc4)c2c1