accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3H3CCH3OOBr

CAS RN 108052-76-2

IUPAC Name: tert-butyl 4-(bromomethyl)benzoate
Synonyms: tert-butyl 4-(bromomethyl)benzoate
Molecular Weight (g/mol): 271.15
Molecular Formula: C12H15BrO2
InChi Key: GSIBTIUXYYFCPU-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)C1=CC=C(CBr)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results