CAS RN 1080533-15-8
CAS RN 1080533-15-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Mirtazapine N-Glucuronide (Mixture of Diastereomers) Contains Unknown Inorganics >80%, TRC
CAS: 1080533-15-8 Molecular Formula: C23 H28 N3 O6 Molecular Weight (g/mol): 442.48 Synonym: 2-β-D-Glucopyranuronosyl-1,2,3,4,10,14b-hexahydro-2-methyl-pyrazino[2,1-a]pyrido[2,3-c][2]benzazepinium SMILES: C[N+]1(CCN2C(C1)c3ccccc3Cc4cccnc24)[C@@H]5O[C@@H]([C@@H](O)[C@H](O)[C@H]5O)C(=O)O