accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 108087-84-9

CH3OHNOOO(Z)

CAS RN 108087-84-9

IUPAC Name: (2Z)-3-[(4-ethoxyphenyl)carbamoyl]prop-2-enoate
Synonyms: (2Z)-3-[(4-ethoxyphenyl)carbamoyl]prop-2-enoate
Molecular Weight (g/mol): 234.23
Molecular Formula: C12H12NO4
InChi Key: LMTIUWRMRAZPEN-FPLPWBNLSA-M
SMILES: CCOC1=CC=C(NC(=O)\C=C/C([O-])=O)C=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results