accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

ONONHNCH2NOO

CAS RN 1081-15-8

IUPAC Name: 1-(2,4-dinitrophenyl)-2-methylidenehydrazine
Synonyms: 1-(2,4-dinitrophenyl)-2-methylidenehydrazine
Molecular Weight (g/mol): 210.15
Molecular Formula: C7H6N4O4
InChi Key: UEQLSLWCHGLSML-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=C(NN=C)C(=C1)[N+]([O-])=O
Quantity 
  • (1)
  • (1)
Concentration 
  • (1)
Solvent or Matrix 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results