accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 1081-75-0

IUPAC Name: (3-phenylpropyl)benzene
Synonyms: 1,3-diphenylpropane propane,1,3-diphenyl
Molecular Weight (g/mol): 196.29
Molecular Formula: C15H16
InChi Key: VEAFKIYNHVBNIP-UHFFFAOYSA-N
SMILES: C(CC1=CC=CC=C1)CC1=CC=CC=C1
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)