accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3C

CAS RN 1081-77-2

IUPAC Name: nonylbenzene
Synonyms: N-nonylbenzene
Molecular Weight (g/mol): 204.36
Molecular Formula: C15H24
InChi Key: LIXVMPBOGDCSRM-UHFFFAOYSA-N
SMILES: CCCCCCCCCC1=CC=CC=C1
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results