CAS RN 1083100-26-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
N-[3-Methyl-4-(2,2,2-trifluoroethoxy)-2-pyridinyl]methyl Lansoprazole, TRC
CAS: 1083100-26-8 Molecular Formula: C25H22F6N4O3S Molecular Weight (g/mol): 572.52 Synonym: 1-[[3-Methyl-4-(2,2,2-trifluoroethoxy)-2-pyridinyl]methyl]-2-[[[3-methyl-4-(2,2,2-trifluoroethoxy)-2-pyridinyl]methyl]sulfinyl]-1H-benzimidazole,Lansoprazole Impurity IUPAC Name: 1-[[3-methyl-4-(2,2,2-trifluoroethoxy)pyridin-2-yl]methyl]-2-[[3-methyl-4-(2,2,2-trifluoroethoxy)pyridin-2-yl]methylsulfinyl]benzimidazole SMILES: Cc1c(OCC(F)(F)F)ccnc1CS(=O)c2nc3ccccc3n2Cc4nccc(OCC(F)(F)F)c4C