accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

(S)O(R)(R)(S)(R)(R)(S)(S)(R)(R)O(R)O(R)(R)(S)ONH2NHHOHOHOHOH2NNH2OHHOCH3H3CCH3SOOOHOHSOOOHOH

CAS RN 108321-42-2

IUPAC Name: 5-amino-6-[(4,6-diamino-3-{[3,5-dihydroxy-5-methyl-4-(methylamino)oxan-2-yl]oxy}-2-hydroxycyclohexyl)oxy]-2-(1-hydroxyethyl)oxane-3,4-diol; bis(sulfuric acid)
Synonyms: 5-amino-6-[(4,6-diamino-3-{[3,5-dihydroxy-5-methyl-4-(methylamino)oxan-2-yl]oxy}-2-hydroxycyclohexyl)oxy]-2-(1-hydroxyethyl)oxane-3,4-diol; bis(sulfuric acid)
Molecular Weight (g/mol): 692.7
Molecular Formula: C20H44N4O18S2
InChi Key: UHEPSJJJMTWUCP-UHFFFAOYNA-N
SMILES: OS(O)(=O)=O.OS(O)(=O)=O.CNC1C(O)C(OC2C(N)CC(N)C(OC3OC(C(C)O)C(O)C(O)C3N)C2O)OCC1(C)O
Quantity 
  • (4)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (5)
  • (3)
Molecular Weight (g/mol) 
  • (9)
  • (2)
Physical Form 
  • (4)
  • (5)
  • (2)
Type 
  • (5)
  • (6)
Color 
  • (2)
  • (7)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (6)
  • (5)
  • (1)
  • (2)
  • (5)
  • (1)
  • (2)
  • (2)
  • (1)
  • (4)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (5)
  • (3)
  • (9)
  • (2)
  • (4)
  • (5)
  • (2)
  • (5)
  • (6)
  • (2)
  • (7)
  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (3)
  • (3)