accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 1086375-50-9

IUPAC Name: pyrazolo[1,5-a]pyrimidine-5-carboxylic acid
Synonyms: pyrazolo[1,5-a]pyrimidine-5-carboxylic acid
Molecular Weight (g/mol): 163.14
Molecular Formula: C7H5N3O2
InChi Key: SBDWLDBGGVPVAR-UHFFFAOYSA-N
SMILES: OC(=O)C1=NC2=CC=NN2C=C1
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results