accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3NOBr

CAS RN 1086386-20-0

IUPAC Name: 7-bromo-1-methyl-1,2,3,4-tetrahydroquinolin-2-one
Synonyms: 7-bromo-1-methyl-3,4-dihydroquinolin-2-one
Molecular Weight (g/mol): 240.1
Molecular Formula: C10H10BrNO
InChi Key: XVBPEXCFWANWIM-UHFFFAOYSA-N
SMILES: CN1C(=O)CCC2=C1C=C(Br)C=C2

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results