CAS RN 108656-33-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Florfenicol Amine Hydrochloride, TRC
CAS: 108656-33-3 Molecular Formula: C10 H14 F N O3 S . Cl H Molecular Weight (g/mol): 283.75 Synonym: Florfenicol Amine Hydrochloride,Benzenemethanol, α-(1-amino-2-fluoroethyl)-4-(methylsulfonyl)-, hydrochloride, [R-(R*,S*)]- (9CI),Florfenicol-amine hydrochloride IUPAC Name: (1R,2S)-2-amino-3-fluoro-1-(4-methylsulfonylphenyl)propan-1-ol;hydrochloride SMILES: Cl.CS(=O)(=O)c1ccc(cc1)[C@@H](O)[C@H](N)CF