accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

HOBOHSS

CAS RN 108847-76-3

IUPAC Name: (thianthren-1-yl)boronic acid
Synonyms: thianthren-1-ylboronic acid
Molecular Weight (g/mol): 260.13
Molecular Formula: C12H9BO2S2
InChi Key: FZEWPLIHPXGNTB-UHFFFAOYSA-N
SMILES: OB(O)C1=CC=CC2=C1SC1=CC=CC=C1S2
Grade 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results