accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NH2NN

CAS RN 109-12-6

IUPAC Name: pyrimidin-2-amine
Synonyms: 2-aminopyrimidine 2-pyridiylamine 2-pyrimidiylamine
Molecular Weight (g/mol): 95.11
Molecular Formula: C4H5N3
InChi Key: LJXQPZWIHJMPQQ-UHFFFAOYSA-N
SMILES: NC1=NC=CC=N1
Molecular Weight (g/mol) 
  • (3)
  • (5)
Quantity 
  • (2)
  • (2)
  • (2)
Boiling Point 
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results