accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3COOCH3

CAS RN 109-21-7

IUPAC Name: butyl butanoate
Synonyms: butyl butyrate N-butylbutyrate
Molecular Weight (g/mol): 144.21
Molecular Formula: C8H16O2
InChi Key: XUPYJHCZDLZNFP-UHFFFAOYSA-N
SMILES: CCCCOC(=O)CCC
Boiling Point 
  • (2)
  • (3)
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (2)
  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)