accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OO

CAS RN 109-29-5

IUPAC Name: 1-oxacycloheptadecan-2-one
Synonyms: hexadecanolactone
Molecular Weight (g/mol): 254.41
Molecular Formula: C16H30O2
InChi Key: LOKPJYNMYCVCRM-UHFFFAOYSA-N
SMILES: O=C1CCCCCCCCCCCCCCCO1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results