accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3COCH2

CAS RN 109-49-9

IUPAC Name: hex-5-en-2-one
Synonyms: 1-hexen-5-one
Molecular Weight (g/mol): 98.15
Molecular Formula: C6H10O
InChi Key: RNDVGJZUHCKENF-UHFFFAOYSA-N
SMILES: CC(=O)CCC=C
Boiling Point 
  • (2)
  • (3)
Molecular Weight (g/mol) 
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (2)
  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)