accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3COHO

CAS RN 109-52-4

IUPAC Name: pentanoic acid
Synonyms: N-valeric acid kyselina valerova ammonium salt n-pentanoic acid valeriansaeure n-pentanoic acid, 11C-labeled 11C-labeled n-pentanoic acid n-BuCOOH zinc salt n-pentanoic acid valeric acid normal valeriansaure
Molecular Weight (g/mol): 102.13
Molecular Formula: C5H10O2
InChi Key: NQPDZGIKBAWPEJ-UHFFFAOYSA-N
SMILES: CCCCC(O)=O
Molecular Weight (g/mol) 
  • (4)
  • (8)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (3)
  • (4)
  • (3)
  • (4)
  • (8)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)