accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 109-57-9

IUPAC Name: (prop-2-en-1-yl)thiourea
Synonyms: allylthiourea
Molecular Weight (g/mol): 116.18
Molecular Formula: C4H8N2S
InChi Key: HTKFORQRBXIQHD-UHFFFAOYSA-N
SMILES: NC(=S)NCC=C
Quantity 
  • (2)
  • (3)
  • (1)
  • (2)
  • (1)
Molecular Weight (g/mol) 
  • (3)
  • (2)
  • (3)
Physical Form 
  • (3)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (1)
  • (2)
  • (1)
  • (3)
  • (2)
  • (3)
  • (3)
  • (1)