accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3COCH3O

CAS RN 109-60-4

IUPAC Name: propyl acetate
Synonyms: propyl acetate octan propylu propyl acetate, ion (1-) ion (1-) propyl acetate N-propyl acetate propylester kyseliny octove n-propyl acetate (natural) acetate de propyle normal propyl acetate (substance)
Molecular Weight (g/mol): 102.13
Molecular Formula: C5H10O2
InChi Key: YKYONYBAUNKHLG-UHFFFAOYSA-N
SMILES: CCCOC(C)=O
Boiling Point 
  • (4)
  • (3)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (5)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (4)
  • (3)
  • (2)
  • (2)
  • (1)
  • (1)
  • (3)
  • (2)
  • (2)
  • (5)
  • (5)
  • (1)
  • (3)
  • (1)