accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 109-67-1

IUPAC Name: pent-1-ene
Synonyms: pentene
Molecular Weight (g/mol): 70.14
Molecular Formula: C5H10
InChi Key: YWAKXRMUMFPDSH-UHFFFAOYSA-N
SMILES: CCCC=C
Boiling Point 
  • (2)
  • (5)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (5)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (5)
  • (1)
  • (1)