CAS RN 109-68-2
IUPAC Name:
(2Z)-pent-2-ene
Synonyms:
cis-2-pentene
Molecular Weight (g/mol):
70.14
Molecular Formula:
C5H10
InChi Key:
QMMOXUPEWRXHJS-HYXAFXHYSA-N
SMILES:
CC\C=C/C
1
–
1
of
1
results
1
2-Pentene, cis + trans, 98%
CAS: 109-68-2 Molecular Formula: C5H10 Molecular Weight (g/mol): 70.14 MDL Number: MFCD00009384 InChI Key: QMMOXUPEWRXHJS-HYXAFXHYSA-N Synonym: trans-2-pentene,e-2-pentene,2-pentene,3-pentene,2-pentene, e,sym-methylethylethylene,e-pent-2-ene,beta-n-amylene,trans-beta-amylene,2-pentene, 2e PubChem CID: 5326161 SMILES: CC\C=C/C