accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

ClBr

CAS RN 109-70-6

IUPAC Name: 1-bromo-3-chloropropane
Synonyms: 1-chloro-3-bromopropane
Molecular Weight (g/mol): 157.44
Molecular Formula: C3H6BrCl
InChi Key: MFESCIUQSIBMSM-UHFFFAOYSA-N
SMILES: ClCCCBr
Molecular Weight (g/mol) 
  • (5)
  • (5)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (5)
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (5)
  • (5)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (5)
  • (4)