accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CLi

CAS RN 109-72-8

IUPAC Name: lithium(1+) butan-1-ide
Synonyms: N-butyllithium
Molecular Weight (g/mol): 64.06
Molecular Formula: C4H9Li
InChi Key: DLEDOFVPSDKWEF-UHFFFAOYSA-N
SMILES: [Li+].CCC[CH2-]
Quantity 
  • (6)
  • (2)
  • (3)
  • (5)
Physical Form 
  • (8)
  • (3)
Boiling Point 
  • (6)
Percent Purity 
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (7)
  • (8)
  • (6)
  • (2)
  • (3)
  • (5)
  • (8)
  • (3)
  • (6)
  • (3)
  • (2)