accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3H2N

CAS RN 109-73-9

IUPAC Name: butan-1-amine
Synonyms: butylamine fema number 3130 n-butilamina monobutilamina norvalamine n-C4H9NH2
Molecular Weight (g/mol): 73.14
Molecular Formula: C4H11N
InChi Key: HQABUPZFAYXKJW-UHFFFAOYSA-N
SMILES: CCCCN
Boiling Point 
  • (3)
  • (1)
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
Molecular Weight (g/mol) 
  • (4)
  • (3)
Percent Purity 
  • (3)
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (1)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (4)
  • (3)
  • (3)
  • (4)
  • (5)