accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NH2H2N

CAS RN 109-76-2

IUPAC Name: propane-1,3-diamine
Synonyms: α,ω-propanediamine tmeda a,w-propanediamine
Molecular Weight (g/mol): 74.13
Molecular Formula: C3H10N2
InChi Key: XFNJVJPLKCPIBV-UHFFFAOYSA-N
SMILES: NCCCN
Molecular Weight (g/mol) 
  • (1)
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (3)
  • (3)
Percent Purity 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)