accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NN

CAS RN 109-77-3

IUPAC Name: propanedinitrile
Synonyms: malononitrile malonsaeuredinitril usaf KF-19 dwumetylosulfotlenku malonodinitrile malonic dinitrile methylene cyanide methylenecyanide nitrile kyseliny malonove methylenedinitrile
Molecular Weight (g/mol): 66.06
Molecular Formula: C3H2N2
InChi Key: CUONGYYJJVDODC-UHFFFAOYSA-N
SMILES: N#CCC#N
Molecular Weight (g/mol) 
  • (7)
  • (3)
Quantity 
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
Boiling Point 
  • (4)
  • (3)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (4)
  • (3)
  • (7)
  • (3)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (4)
  • (3)
  • (3)