CAS RN 109-88-6
CAS RN 109-88-6
IUPAC Name:
magnesium(2+) dimethanolate
Synonyms:
magnesium(2+) bis(methoxide)
Molecular Weight (g/mol):
86.37
Molecular Formula:
C2H6MgO2
InChi Key:
CRGZYKWWYNQGEC-UHFFFAOYSA-N
SMILES:
[Mg++].C[O-].C[O-]